Product Details
- SNP ID
-
rs4899476
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:73718236 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AGTACCTGCCGCGGCCTCCCTCCCA[A/G]GGAGGAGGTGGCCACTCAGGACACG
- Phenotype
-
MIM: 604010
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ELMSAN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111854480] are located under a probe and SNP(s) [rs60510049] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ELMSAN1
- Gene Name
- ELM2 and Myb/SANT domain containing 1
- Gene
- PNMA1
- Gene Name
- paraneoplastic Ma antigen 1
There are no transcripts associated with this gene.
View Full Product Details