Product Details

SNP ID
rs7145768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:37217514 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTCAACAAAATACTAAAACCGA[A/G]TTCAGCAATCCATTTGAAAGATCAT
Phenotype
MIM: 606850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIPOL1 PubMed Links

Gene Details

Gene
MIPOL1
Gene Name
mirror-image polydactyly 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195296.1 Intron NP_001182225.1
NM_001195297.1 Intron NP_001182226.1
NM_138731.6 Intron NP_620059.1
XM_011536463.2 Intron XP_011534765.1
XM_011536467.2 Intron XP_011534769.1
XM_017020991.1 Intron XP_016876480.1
XM_017020992.1 Intron XP_016876481.1
XM_017020993.1 Intron XP_016876482.1
XM_017020994.1 Intron XP_016876483.1
XM_017020995.1 Intron XP_016876484.1
XM_017020996.1 Intron XP_016876485.1
XM_017020997.1 Intron XP_016876486.1
XM_017020998.1 Intron XP_016876487.1
XM_017020999.1 Intron XP_016876488.1
XM_017021000.1 Intron XP_016876489.1
XM_017021001.1 Intron XP_016876490.1
XM_017021002.1 Intron XP_016876491.1
XM_017021003.1 Intron XP_016876492.1
XM_017021004.1 Intron XP_016876493.1
XM_017021005.1 Intron XP_016876494.1
XM_017021006.1 Intron XP_016876495.1
XM_017021007.1 Intron XP_016876496.1
XM_017021008.1 Intron XP_016876497.1
XM_017021009.1 Intron XP_016876498.1
XM_017021010.1 Intron XP_016876499.1
XM_017021011.1 Intron XP_016876500.1
XM_017021012.1 Intron XP_016876501.1
XM_017021013.1 Intron XP_016876502.1
XM_017021014.1 Intron XP_016876503.1
XM_017021015.1 Intron XP_016876504.1
XM_017021016.1 Intron XP_016876505.1
XM_017021017.1 Intron XP_016876506.1
XM_017021018.1 Intron XP_016876507.1
XM_017021019.1 Intron XP_016876508.1

View Full Product Details