Product Details
- SNP ID
-
rs12671270
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:26646315 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCATTACAGAGCTTTCTCCGCATG[C/T]GGGACTCTCGGCCTCATTATGAGTG
- Phenotype
-
MIM: 605215
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C7orf71
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114775093] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C7orf71
- Gene Name
- chromosome 7 open reading frame 71
- Gene
- SKAP2
- Gene Name
- src kinase associated phosphoprotein 2
There are no transcripts associated with this gene.
View Full Product Details