Product Details
- SNP ID
-
rs8051196
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:10947512 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGAATGAACAGAAGCCACCATTGG[C/G]GGGCAGCAGGGGCGGGAGGCGGAGC
- Phenotype
-
MIM: 600005
MIM: 611303
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CIITA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115418449] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CIITA
- Gene Name
- class II, major histocompatibility complex, transactivator
There are no transcripts associated with this gene.
- Gene
- CLEC16A
- Gene Name
- C-type lectin domain family 16 member A
- Gene
- DEXI
- Gene Name
- Dexi homolog
There are no transcripts associated with this gene.
View Full Product Details