Product Details

SNP ID
rs13270615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33314109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTAAAGTGCTGAGTTATCATC[A/G]GCCTATTTTTTCCCTGCTAATGTTT
Phenotype
MIM: 616931
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FUT10 PubMed Links

Gene Details

Gene
FUT10
Gene Name
fucosyltransferase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032664.3 Intron NP_116053.3
XM_011544676.2 Intron XP_011542978.1
XM_011544677.2 Intron XP_011542979.1
XM_011544678.2 Intron XP_011542980.1
XM_011544679.2 Intron XP_011542981.1
XM_011544680.2 Intron XP_011542982.1
XM_011544681.2 Intron XP_011542983.1

View Full Product Details