Product Details

SNP ID
rs12578773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:25216093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAACTGGAACCTTGAGTTTTAT[C/T]TTAAATTTTAACACCTTTCAGTCTA
Phenotype
MIM: 190070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRAS PubMed Links

Gene Details

Gene
KRAS
Gene Name
KRAS proto-oncogene, GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004985.4 Intron NP_004976.2
NM_033360.3 Intron NP_203524.1
XM_006719069.3 Intron XP_006719132.1
XM_011520653.2 Intron XP_011518955.1
XM_017019292.1 Intron XP_016874781.1
XM_017019293.1 Intron XP_016874782.1

View Full Product Details