Product Details

SNP ID
rs3798420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118465323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAATCCCTAAGTGCAAGTCATG[C/T]CACTTGCAACCTTCCATTATGGCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP85L PubMed Links

Gene Details

Gene
CEP85L
Gene Name
centrosomal protein 85 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042475.2 3679 UTR 3 NP_001035940.1
NM_001178035.1 3679 UTR 3 NP_001171506.1
NM_206921.2 3679 Intron NP_996804.2
XM_005266970.1 3679 UTR 3 XP_005267027.1
XM_005266971.1 3679 UTR 3 XP_005267028.1
XM_005266972.4 3679 UTR 3 XP_005267029.1
XM_006715475.3 3679 UTR 3 XP_006715538.1
XM_011535809.2 3679 UTR 3 XP_011534111.1
XM_011535810.2 3679 UTR 3 XP_011534112.1
XM_011535811.1 3679 UTR 3 XP_011534113.1
XM_017010846.1 3679 UTR 3 XP_016866335.1
XM_017010847.1 3679 UTR 3 XP_016866336.1

View Full Product Details