Product Details
- SNP ID
-
rs7430680
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:46701259 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGGGGATGCGGAAGGTGGGGGC[A/T]GGGAGGGGGAGCCTGCGGCCCGATC
- Phenotype
-
MIM: 612402
MIM: 607237
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ALS2CL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs184330048,rs192733532] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALS2CL
- Gene Name
- ALS2 C-terminal like
There are no transcripts associated with this gene.
- Gene
- TMIE
- Gene Name
- transmembrane inner ear
View Full Product Details