Product Details
- SNP ID
-
rs1538369
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
28
- Location
-
Chr.1:205506873 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAGAGGGCACCAGATCTGAGGAGC[A/T]AATAGGTTCTTGGTCTGAGATCTCG
- Phenotype
-
MIM: 169190
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDK18
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75049559] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDK18
- Gene Name
- cyclin dependent kinase 18
View Full Product Details