Product Details

SNP ID
rs6112852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2100945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTTCAGCGACCAAATACATTAT[A/G]AGAAATGGCCTGTATCCAGTGAGCA
Phenotype
MIM: 609370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STK35 PubMed Links

Gene Details

Gene
STK35
Gene Name
serine/threonine kinase 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080836.3 2911 Intron NP_543026.2
XM_011529174.2 2911 UTR 5 XP_011527476.1

View Full Product Details