Product Details

SNP ID
rs4339500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110190611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACATTCATATAATGAAATTTAGTA[A/G]GCAGTTGGATATGTGGATCCAACTG
Phenotype
MIM: 605585 MIM: 605035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC40 PubMed Links

Gene Details

Gene
CDC40
Gene Name
cell division cycle 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015891.2 Intron NP_056975.1
Gene
WASF1
Gene Name
WAS protein family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024934.1 Intron NP_001020105.1
NM_001024935.1 Intron NP_001020106.1
NM_001024936.1 Intron NP_001020107.1
NM_003931.2 Intron NP_003922.1
XM_005267203.3 Intron XP_005267260.1
XM_005267205.2 Intron XP_005267262.1
XM_005267206.2 Intron XP_005267263.1
XM_005267207.2 Intron XP_005267264.1
XM_011536233.1 Intron XP_011534535.1
XM_011536234.1 Intron XP_011534536.1
XM_011536235.1 Intron XP_011534537.1
XM_017011437.1 Intron XP_016866926.1
XM_017011438.1 Intron XP_016866927.1
XM_017011439.1 Intron XP_016866928.1
XM_017011440.1 Intron XP_016866929.1
XM_017011441.1 Intron XP_016866930.1
XM_017011442.1 Intron XP_016866931.1
XM_017011443.1 Intron XP_016866932.1
XM_017011444.1 Intron XP_016866933.1

View Full Product Details