Product Details

SNP ID
rs34941234
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:23135595 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATAAACGAACACCACACCTTTTTAA[C/T]ATGAGGTTTCATATTTATTTTGGTC
Phenotype
MIM: 603614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF10D PubMed Links

Gene Details

Gene
TNFRSF10D
Gene Name
TNF receptor superfamily member 10d
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003840.4 3531 UTR 3 NP_003831.2

View Full Product Details