Product Details

SNP ID
rs2790444
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:28536760 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAACTATTTTCAGACTTAATTTGCC[C/T]GAATTACATTTCATGTCAAAAGCAT
Phenotype
MIM: 615049
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WAC PubMed Links

Gene Details

Gene
WAC
Gene Name
WW domain containing adaptor with coiled-coil
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016628.4 Intron NP_057712.2
NM_100264.2 Intron NP_567822.1
NM_100486.3 Intron NP_567823.1
XM_011519491.2 Intron XP_011517793.1
XM_017016315.1 Intron XP_016871804.1
XM_017016316.1 Intron XP_016871805.1
XM_017016317.1 Intron XP_016871806.1
XM_017016318.1 Intron XP_016871807.1
Gene
WAC-AS1
Gene Name
WAC antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details