Product Details

SNP ID
rs12437095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:38209151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACATCCTGGCCTCTCCTCTCCAC[A/G]GTCGCCTGTGCCCGGGCACCCCGGA
Phenotype
MIM: 182451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SSTR1 PubMed Links

Gene Details

Gene
SSTR1
Gene Name
somatostatin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001049.2 379 UTR 5 NP_001040.1

View Full Product Details