Product Details

SNP ID
rs7646039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114357288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAAATTTGCAGAAATTTGACTT[C/T]GAACTTGCAGAAATTTGACTTTCAA
Phenotype
MIM: 606025
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB20 PubMed Links

Gene Details

Gene
ZBTB20
Gene Name
zinc finger and BTB domain containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164342.2 Intron NP_001157814.1
NM_001164343.2 Intron NP_001157815.1
NM_001164344.2 Intron NP_001157816.1
NM_001164345.2 Intron NP_001157817.1
NM_001164346.2 Intron NP_001157818.1
NM_001164347.2 Intron NP_001157819.1
NM_015642.5 Intron NP_056457.3
Gene
ZBTB20-AS1
Gene Name
ZBTB20 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details