Product Details

SNP ID
rs7493080
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:69271362 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGGCCAGCGCTGGGGCAAAGCTGA[A/G]GGCAGGCCAACCAGAGAGCCCAAGA
Phenotype
MIM: 615132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALNT16 PubMed Links

Gene Details

Gene
GALNT16
Gene Name
polypeptide N-acetylgalactosaminyltransferase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168368.1 Intron NP_001161840.1
NM_020692.2 Intron NP_065743.2
XM_011537005.1 Intron XP_011535307.1
XM_011537006.2 Intron XP_011535308.1
XM_011537007.2 Intron XP_011535309.1
XM_017021498.1 Intron XP_016876987.1

View Full Product Details