Product Details

SNP ID
rs11882639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44475380 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCCTTAATATTGCTTTGCATATA[C/T]GAAAATAAGCCCACTTGCCCTCTAT
Phenotype
MIM: 606740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF180 PubMed Links

Gene Details

Gene
ZNF180
Gene Name
zinc finger protein 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278508.2 3262 UTR 3 NP_001265437.2
NM_001278509.2 3262 UTR 3 NP_001265438.2
NM_001288759.2 3262 UTR 3 NP_001275688.2
NM_001288760.2 3262 UTR 3 NP_001275689.1
NM_001288761.2 3262 UTR 3 NP_001275690.1
NM_001288762.2 3262 UTR 3 NP_001275691.1
NM_001291633.1 3262 UTR 3 NP_001278562.1
NM_013256.5 3262 UTR 3 NP_037388.3
XM_011527280.2 3262 UTR 3 XP_011525582.1
XM_017027250.1 3262 UTR 3 XP_016882739.1
XM_017027251.1 3262 UTR 3 XP_016882740.1
XM_017027252.1 3262 UTR 3 XP_016882741.1
XM_017027253.1 3262 UTR 3 XP_016882742.1
XM_017027254.1 3262 UTR 3 XP_016882743.1
XM_017027255.1 3262 UTR 3 XP_016882744.1
XM_017027256.1 3262 UTR 3 XP_016882745.1

View Full Product Details