Product Details

SNP ID
rs2571051
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:44475226 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCCTCATGAAAGCATTTTTGCAAG[C/T]ATAATTTAGACGGTCAGCACTGAGC
Phenotype
MIM: 606740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF180 PubMed Links

Gene Details

Gene
ZNF180
Gene Name
zinc finger protein 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278508.2 3416 UTR 3 NP_001265437.2
NM_001278509.2 3416 UTR 3 NP_001265438.2
NM_001288759.2 3416 UTR 3 NP_001275688.2
NM_001288760.2 3416 UTR 3 NP_001275689.1
NM_001288761.2 3416 UTR 3 NP_001275690.1
NM_001288762.2 3416 UTR 3 NP_001275691.1
NM_001291633.1 3416 UTR 3 NP_001278562.1
NM_013256.5 3416 UTR 3 NP_037388.3
XM_011527280.2 3416 UTR 3 XP_011525582.1
XM_017027250.1 3416 UTR 3 XP_016882739.1
XM_017027251.1 3416 UTR 3 XP_016882740.1
XM_017027252.1 3416 UTR 3 XP_016882741.1
XM_017027253.1 3416 UTR 3 XP_016882742.1
XM_017027254.1 3416 UTR 3 XP_016882743.1
XM_017027255.1 3416 UTR 3 XP_016882744.1
XM_017027256.1 3416 UTR 3 XP_016882745.1

View Full Product Details