Product Details

SNP ID
rs34542537
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:18840007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCATCGGGGTCAGTACGGGTGGT[A/G]GGACTGAGGCCCGTGGCCAGCATGC
Phenotype
MIM: 606226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAS1R2 PubMed Links

Gene Details

Gene
TAS1R2
Gene Name
taste 1 receptor member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152232.2 2113 Silent Mutation CCC,CCT P704P NP_689418.2

View Full Product Details