Product Details

SNP ID
rs2233788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177992586 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGATCTCCCATTTTTAAATTTCTA[A/G]TCGCTGAGCTGACCCTCACCATGCA
Phenotype
MIM: 601538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PROP1 PubMed Links

Gene Details

Gene
PROP1
Gene Name
PROP paired-like homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006261.4 1113 UTR 3 NP_006252.3

View Full Product Details