Product Details
- SNP ID
-
rs7905015
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:75103332 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAGAAGACTCAGGCTGAGGGAAGC[A/G]GCACAGGCCCACACACAGACACAGG
- Phenotype
-
MIM: 613191
MIM: 611575
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DUSP13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145950091] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DUSP13
- Gene Name
- dual specificity phosphatase 13
- Gene
- SAMD8
- Gene Name
- sterile alpha motif domain containing 8
View Full Product Details