Product Details

SNP ID
rs2001823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240573053 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTGCCTGACAGGCTCCCCGGCC[G/T]GGGCTATGGGTGTGCTGACGGTGGG
Phenotype
MIM: 605287
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CAPN10-AS1 PubMed Links

Gene Details

Gene
CAPN10-AS1
Gene Name
CAPN10 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
DUSP28
Gene Name
dual specificity phosphatase 28
There are no transcripts associated with this gene.

Gene
RNPEPL1
Gene Name
arginyl aminopeptidase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018226.4 Intron NP_060696.4
XM_005247036.4 Intron XP_005247093.1

View Full Product Details