Product Details

SNP ID
rs11609532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:82694586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGACATGGTATGAAGTGTGGCAC[A/G]GCATGTAGTTATAGTCAAGATGACT
Phenotype
MIM: 615856
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMTC2 PubMed Links

Gene Details

Gene
TMTC2
Gene Name
transmembrane and tetratricopeptide repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320321.1 Intron NP_001307250.1
NM_001320322.1 Intron NP_001307251.1
NM_152588.2 Intron NP_689801.1
XM_017018884.1 Intron XP_016874373.1

View Full Product Details