Product Details

SNP ID
rs12569041
Assay Type
Functionally Tested
NCBI dbSNP Submissions
47
Location
Chr.1:118884831 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCCTGTTGGTTATTGGGTGAAGG[A/G]CTAATCATGTGCATGCTGTGCTCCA
Phenotype
MIM: 604127
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TBX15 PubMed Links

Gene Details

Gene
TBX15
Gene Name
T-box 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152380.2 2163 Silent Mutation AGC,AGT S464S NP_689593.2
XM_005271161.3 2163 Silent Mutation AGC,AGT S603S XP_005271218.1
XM_005271162.1 2163 Silent Mutation AGC,AGT S570S XP_005271219.1
XM_011542052.2 2163 Silent Mutation AGC,AGT S288S XP_011540354.1

View Full Product Details