Product Details

SNP ID
rs35757182
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:62640357 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTCCCAGGAGGTCACGGTGGAGAGT[C/G]AGTGGCTAGAGGCAGAGAGAAGAGG
Phenotype
MIM: 607349
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MGC15885 PubMed Links

Gene Details

Gene
MGC15885
Gene Name
uncharacterized protein MGC15885
There are no transcripts associated with this gene.

Gene
TLN2
Gene Name
talin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015059.2 Intron NP_055874.2
XM_005254708.4 Intron XP_005254765.1
XM_005254710.4 Intron XP_005254767.1
XM_005254711.4 Intron XP_005254768.1
XM_005254712.4 Intron XP_005254769.1
XM_005254713.4 Intron XP_005254770.1
XM_005254714.3 Intron XP_005254771.1
XM_005254715.2 Intron XP_005254772.1
XM_006720717.3 Intron XP_006720780.1
XM_017022665.1 Intron XP_016878154.1
XM_017022666.1 Intron XP_016878155.1
XM_017022667.1 Intron XP_016878156.1
XM_017022668.1 Intron XP_016878157.1
XM_017022669.1 Intron XP_016878158.1

View Full Product Details