Product Details

SNP ID
rs12477095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:191834973 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTGAAGATTTATATTCGTTCCA[A/G]TGTTAACAAGACAACATCCTTTTGA
Phenotype
MIM: 606728
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDPR PubMed Links

Gene Details

Gene
SDPR
Gene Name
serum deprivation response
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004657.5 2583 UTR 3 NP_004648.1

View Full Product Details