Product Details

SNP ID
rs34814776
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:33151100 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CACATACTCATCTGTGCCTTTAGGG[C/T]TTCAGCTGGCCTCGAAGTTCACATT
Phenotype
MIM: 605497
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRTAP PubMed Links

Gene Details

Gene
CRTAP
Gene Name
cartilage associated protein
There are no transcripts associated with this gene.

Gene
SUSD5
Gene Name
sushi domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015551.1 3950 UTR 3 NP_056366.1
XM_005265034.3 3950 UTR 3 XP_005265091.1
XM_017006137.1 3950 UTR 3 XP_016861626.1

View Full Product Details