Product Details

SNP ID
rs12602885
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:80545369 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCCTCAGGAGCAGCCAGTGGCTTG[A/G]GACCTGGGGTGGTGTGTGTCTGCGG
Phenotype
MIM: 607130
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPTOR PubMed Links

Gene Details

Gene
RPTOR
Gene Name
regulatory associated protein of MTOR complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163034.1 545 UTR 5 NP_001156506.1
NM_020761.2 545 UTR 5 NP_065812.1

View Full Product Details