Product Details
- SNP ID
-
rs35217513
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:40515952 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCAGACTGATGGTGGGGGGCCTG[A/C]CAGGGAATGTTTCCCAGCACCTTGA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF620
PubMed Links
Gene Details
- Gene
- ZNF620
- Gene Name
- zinc finger protein 620
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256167.1 |
3688 |
Missense Mutation |
ACA,CCA |
T6P |
NP_001243096.1 |
NM_001256168.1 |
3688 |
Missense Mutation |
ACA,CCA |
T6P |
NP_001243097.1 |
NM_175888.3 |
3688 |
Missense Mutation |
ACA,CCA |
T120P |
NP_787084.1 |
XM_005265011.3 |
3688 |
Missense Mutation |
ACA,CCA |
T120P |
XP_005265068.1 |
XM_005265012.3 |
3688 |
Missense Mutation |
ACA,CCA |
T120P |
XP_005265069.1 |
XM_017006064.1 |
3688 |
Missense Mutation |
ACA,CCA |
T165P |
XP_016861553.1 |
XM_017006065.1 |
3688 |
Missense Mutation |
ACA,CCA |
T82P |
XP_016861554.1 |
XM_017006066.1 |
3688 |
Missense Mutation |
ACA,CCA |
T81P |
XP_016861555.1 |
XM_017006067.1 |
3688 |
Missense Mutation |
ACA,CCA |
T81P |
XP_016861556.1 |
XM_017006068.1 |
3688 |
Missense Mutation |
ACA,CCA |
T43P |
XP_016861557.1 |
XM_017006069.1 |
3688 |
Missense Mutation |
ACA,CCA |
T6P |
XP_016861558.1 |
- Gene
- ZNF621
- Gene Name
- zinc finger protein 621
There are no transcripts associated with this gene.
View Full Product Details