Product Details
- SNP ID
-
rs6651267
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:86523345 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGGAAAGAAATTTGAATGACACAC[A/G]CTGACTCCAGCATTATTGCTCTTTC
- Phenotype
-
MIM: 604207
MIM: 611871
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CPNE3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116579976] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPNE3
- Gene Name
- copine 3
- Gene
- RMDN1
- Gene Name
- regulator of microtubule dynamics 1
There are no transcripts associated with this gene.
View Full Product Details