Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002881.2 | Intron | NP_002872.1 | |||
XM_005263724.1 | Intron | XP_005263781.1 | |||
XM_005263727.1 | Intron | XP_005263784.1 | |||
XM_005263728.1 | Intron | XP_005263785.1 | |||
XM_005263729.2 | Intron | XP_005263786.1 | |||
XM_011511573.1 | Intron | XP_011509875.1 | |||
XM_011511574.1 | Intron | XP_011509876.1 | |||
XM_017004621.1 | Intron | XP_016860110.1 | |||
XM_017004622.1 | Intron | XP_016860111.1 |