Product Details

SNP ID
rs6809
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:64323350 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTGTATTTCCAAAGAACAAGGACTA[G/C]CCAAAACTACAAGCCTTTGAAGGAC
Phenotype
MIM: 173445
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
PECAM1 PubMed Links

Gene Details

Gene
PECAM1
Gene Name
platelet and endothelial cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000442.4 3125 UTR 3 NP_000433.4
XM_005276880.1 3125 UTR 3 XP_005276937.1
XM_005276881.1 3125 UTR 3 XP_005276938.1
XM_005276882.1 3125 UTR 3 XP_005276939.1
XM_005276883.2 3125 Intron XP_005276940.1
XM_011524889.2 3125 UTR 3 XP_011523191.1
XM_011524890.1 3125 UTR 3 XP_011523192.1
XM_017024738.1 3125 UTR 3 XP_016880227.1
XM_017024739.1 3125 UTR 3 XP_016880228.1
XM_017024740.1 3125 UTR 3 XP_016880229.1
XM_017024741.1 3125 UTR 3 XP_016880230.1

View Full Product Details