Product Details

SNP ID
rs35039225
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:28201377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGACCATCTCGACTTTCCTAAGT[A/T]CAAAAGAAGTTTGAGGTGAGGAGAC
Phenotype
MIM: 173393
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC8 PubMed Links

Gene Details

Gene
DNAJC8
Gene Name
DnaJ heat shock protein family (Hsp40) member C8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014280.2 Intron NP_055095.2
Gene
PTAFR
Gene Name
platelet activating factor receptor
There are no transcripts associated with this gene.

View Full Product Details