Product Details

SNP ID
rs35389927
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:45894885 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTGTTTCTGAGGGAGGGCTGGCCCT[A/G]TCTGCTGCACACACTACTGTACATC
Phenotype
MIM: 604641
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPK8IP1 PubMed Links

Gene Details

Gene
MAPK8IP1
Gene Name
mitogen-activated protein kinase 8 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005456.3 Intron NP_005447.1
XM_005253226.3 Intron XP_005253283.1

View Full Product Details