Product Details

SNP ID
rs35108180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:138165092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATATGTTAACATAAACATAAC[A/G]ACACACATATTATTTTTCTACCCCT
Phenotype
MIM: 607933
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC7A11 PubMed Links

Gene Details

Gene
SLC7A11
Gene Name
solute carrier family 7 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014331.3 8650 UTR 3 NP_055146.1
XM_011531802.2 8650 Intron XP_011530104.1
Gene
SLC7A11-AS1
Gene Name
SLC7A11 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details