Product Details

SNP ID
rs10998504
Assay Type
Validated
NCBI dbSNP Submissions
36
Location
Chr.10:68963233 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATGAGTAGTATACTTCAGTATGTC[A/C]GTATGGCTAATGTTTTAAGAATTGC
Phenotype
MIM: 606357 MIM: 610373
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DDX21 PubMed Links

Gene Details

Gene
DDX21
Gene Name
DEAD-box helicase 21
There are no transcripts associated with this gene.

Gene
DDX50
Gene Name
DEAD-box helicase 50
There are no transcripts associated with this gene.

View Full Product Details