Product Details

SNP ID
rs34271523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72368356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTATTGCTCAGTTCGCCATTGC[C/T]TAGAGGAAGAGGCAAAAACAACAGG
Phenotype
MIM: 605084
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MICU1 PubMed Links

Gene Details

Gene
MICU1
Gene Name
mitochondrial calcium uptake 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195518.1 Intron NP_001182447.1
NM_001195519.1 Intron NP_001182448.1
NM_006077.3 Intron NP_006068.2
XM_005269383.2 Intron XP_005269440.1
XM_005269386.2 Intron XP_005269443.1
XM_011539119.1 Intron XP_011537421.1

View Full Product Details