Product Details

SNP ID
rs11871638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41973409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTCCTCCAGGAGGGACCCTCC[C/T]TGGGTCAGCTGTTCCTCAAGAGCCT
Phenotype
MIM: 123830 MIM: 601964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNP PubMed Links

Gene Details

Gene
CNP
Gene Name
2',3'-cyclic nucleotide 3' phosphodiesterase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033133.4 Intron NP_149124.3
XM_006721701.3 Intron XP_006721764.1
XM_011524340.2 Intron XP_011522642.1
Gene
DNAJC7
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
There are no transcripts associated with this gene.

Gene
TTC25
Gene Name
tetratricopeptide repeat domain 25
There are no transcripts associated with this gene.

View Full Product Details