Product Details

SNP ID
rs10882947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97506521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGATATGACATAAAATTTACTTGT[A/G]TGTAACAGCAAATTTTTCAGTCTAT
Phenotype
MIM: 614777 MIM: 616388
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMS19 PubMed Links

Gene Details

Gene
MMS19
Gene Name
MMS19 homolog, cytosolic iron-sulfur assembly component
There are no transcripts associated with this gene.

Gene
UBTD1
Gene Name
ubiquitin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024954.4 Intron NP_079230.1

View Full Product Details