Product Details

SNP ID
rs11870652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80547572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGGAGCTGGGGAGAAAGTTTGC[G/T]GTATCTGTTTTTCGGTGGAGAAAAT
Phenotype
MIM: 607130
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RPTOR PubMed Links

Gene Details

Gene
RPTOR
Gene Name
regulatory associated protein of MTOR complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163034.1 Intron NP_001156506.1
NM_020761.2 Intron NP_065812.1

View Full Product Details