Product Details
- SNP ID
-
rs11867438
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:79777483 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AGAACTCCGGGCAGTTCGTATTACT[A/T]CTTTCTCAGAGTGCTTGGTGGTTTA
- Phenotype
-
MIM: 602770
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CBX2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74851309] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CBX2
- Gene Name
- chromobox 2
View Full Product Details