Product Details

SNP ID
rs11822837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46679391 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCACCAGGAAAGCAGTCAGGAAAC[A/G]AAGCACCTGGTAGTTCTCCTCGGGC
Phenotype
MIM: 602732 MIM: 615088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP1 PubMed Links

Gene Details

Gene
ARHGAP1
Gene Name
Rho GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004308.3 1022 Missense Mutation CGT,TGT R369C NP_004299.1
XM_011520095.1 1022 Missense Mutation CGT,TGT R369C XP_011518397.1
XM_017017720.1 1022 Missense Mutation CGT,TGT R325C XP_016873209.1
Gene
ATG13
Gene Name
autophagy related 13
There are no transcripts associated with this gene.

View Full Product Details