Product Details

SNP ID
rs13172523
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:179740049 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGTCATGTGATACTAAATGACAGTC[A/G]TAGCTTAAGATGCCTTGATTTGAGC
Phenotype
MIM: 114217 MIM: 605424
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CANX PubMed Links

Gene Details

Gene
CANX
Gene Name
calnexin
There are no transcripts associated with this gene.

Gene
MAML1
Gene Name
mastermind like transcriptional coactivator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014757.4 Intron NP_055572.1

View Full Product Details