Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001048183.1 | Intron | NP_001041648.1 | |||
NM_023923.3 | Intron | NP_076412.3 | |||
XM_005245970.2 | Intron | XP_005246027.1 | |||
XM_017002157.1 | Intron | XP_016857646.1 | |||
XM_017002158.1 | Intron | XP_016857647.1 | |||
XM_017002159.1 | Intron | XP_016857648.1 | |||
XM_017002160.1 | Intron | XP_016857649.1 | |||
XM_017002161.1 | Intron | XP_016857650.1 | |||
XM_017002162.1 | Intron | XP_016857651.1 | |||
XM_017002163.1 | Intron | XP_016857652.1 | |||
XM_017002164.1 | Intron | XP_016857653.1 | |||
XM_017002165.1 | Intron | XP_016857654.1 | |||
XM_017002166.1 | Intron | XP_016857655.1 | |||
XM_017002167.1 | Intron | XP_016857656.1 | |||
XM_017002168.1 | Intron | XP_016857657.1 | |||
XM_017002169.1 | Intron | XP_016857658.1 | |||
XM_017002170.1 | Intron | XP_016857659.1 | |||
XM_017002171.1 | Intron | XP_016857660.1 | |||
XM_017002172.1 | Intron | XP_016857661.1 | |||
XM_017002173.1 | Intron | XP_016857662.1 |