Product Details

SNP ID
rs4510829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:99878982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTGGAATGTAACCCACATATAT[A/G]TAGTGCAACTGAAATGCTTATTTAG
Phenotype
MIM: 124090 MIM: 607817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX6C PubMed Links

Gene Details

Gene
COX6C
Gene Name
cytochrome c oxidase subunit 6C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004374.3 Intron NP_004365.1
XM_017013020.1 Intron XP_016868509.1
Gene
VPS13B
Gene Name
vacuolar protein sorting 13 homolog B
There are no transcripts associated with this gene.

View Full Product Details