Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022841.5 | 1291 | Intron | NP_073752.5 | ||
XM_005254603.3 | 1291 | Intron | XP_005254660.2 | ||
XM_011521925.2 | 1291 | Intron | XP_011520227.1 | ||
XM_017022506.1 | 1291 | UTR 5 | XP_016877995.1 | ||
XM_017022507.1 | 1291 | Intron | XP_016877996.1 | ||
XM_017022508.1 | 1291 | Intron | XP_016877997.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286449.1 | 1291 | Intron | NP_001273378.1 | ||
NM_198524.2 | 1291 | Intron | NP_940926.1 | ||
XM_005254359.4 | 1291 | Intron | XP_005254416.1 | ||
XM_005254361.3 | 1291 | Intron | XP_005254418.1 | ||
XM_011521530.2 | 1291 | Intron | XP_011519832.1 | ||
XM_017022160.1 | 1291 | Intron | XP_016877649.1 | ||
XM_017022161.1 | 1291 | Intron | XP_016877650.1 | ||
XM_017022162.1 | 1291 | Intron | XP_016877651.1 | ||
XM_017022163.1 | 1291 | Intron | XP_016877652.1 | ||
XM_017022164.1 | 1291 | Intron | XP_016877653.1 | ||
XM_017022166.1 | 1291 | Intron | XP_016877655.1 | ||
XM_017022167.1 | 1291 | Intron | XP_016877656.1 |