Product Details

SNP ID
rs7181116
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:25675529 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCCCGTGTAACAGCAGCCACACCA[C/T]TGAGAGGCCCTGGAGATGGCTTCCC
Phenotype
MIM: 605855
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP10A PubMed Links

Gene Details

Gene
ATP10A
Gene Name
ATPase phospholipid transporting 10A (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024490.3 Intron NP_077816.1
XM_005268261.4 Intron XP_005268318.1
XM_011521826.2 Intron XP_011520128.1
XM_011521828.2 Intron XP_011520130.1
XM_011521829.2 Intron XP_011520131.1
XM_011521830.2 Intron XP_011520132.2
XM_017022436.1 Intron XP_016877925.1
XM_017022437.1 Intron XP_016877926.1

View Full Product Details