Product Details

SNP ID
rs11651026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59243654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCACCAGGGATCCTTGGCTGTCTC[C/T]TATCTCCATCATCGTACCTATTTTA
Phenotype
MIM: 616317
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GDPD1 PubMed Links

Gene Details

Gene
GDPD1
Gene Name
glycerophosphodiester phosphodiesterase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165993.1 Intron NP_001159465.1
NM_001165994.1 Intron NP_001159466.1
NM_182569.3 Intron NP_872375.2
XM_017024521.1 Intron XP_016880010.1

View Full Product Details