Product Details

SNP ID
rs34034499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81671538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTGAGAACCACTGGGGGACCT[C/G]TGAGGGGCCTGTTGGGGTCAGAGCC
Phenotype
MIM: 614271 MIM: 180073
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL16 PubMed Links

Gene Details

Gene
ARL16
Gene Name
ADP ribosylation factor like GTPase 16
There are no transcripts associated with this gene.

Gene
CCDC137
Gene Name
coiled-coil domain containing 137
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199287.2 Intron NP_954981.1
XM_011524738.2 Intron XP_011523040.1
XM_017024573.1 Intron XP_016880062.1
Gene
OXLD1
Gene Name
oxidoreductase like domain containing 1
There are no transcripts associated with this gene.

Gene
PDE6G
Gene Name
phosphodiesterase 6G
There are no transcripts associated with this gene.

View Full Product Details