Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001261467.1 | 791 | Intron | NP_001248396.1 | ||
NM_032141.3 | 791 | Intron | NP_115517.1 | ||
XM_011525345.2 | 791 | Intron | XP_011523647.1 | ||
XM_017025209.1 | 791 | Intron | XP_016880698.1 | ||
XM_017025210.1 | 791 | UTR 5 | XP_016880699.1 | ||
XM_017025211.1 | 791 | Intron | XP_016880700.1 | ||
XM_017025212.1 | 791 | Intron | XP_016880701.1 | ||
XM_017025213.1 | 791 | Intron | XP_016880702.1 | ||
XM_017025214.1 | 791 | Intron | XP_016880703.1 | ||
XM_017025215.1 | 791 | Intron | XP_016880704.1 |